Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sample loops for PerkinElmer
Sample loops for PerkinElmer
Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400, 200 µl
Catalog # HEWL8003-0679
Supplier:   Agilent
Sample loops for PerkinElmer
Catalog # HEWL8003-0679
Supplier:  Agilent
Sample loops for PerkinElmer
Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400, 200 µl
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These sample loops are fully compatible with your PerkinElmer FIMS/FIAS systems, so can be confident that they will integrate seamlessly with PerkinElmer AA system.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure to get the best performance from PerkinElmer system when using parts and supplies.
Specifications
  • Capacity:
    200 µl
  • Description:
    Sample loop, for FIMS-100, FIMS-400, FIAS-100, or FIAS-400
  • Seq:
    01