Inoculating loops, PS
Aseptic, 5 µl, 20/microseal pouch, 200 mm
Catalog # 612-5768
Supplier: Local
Jump to:
Product Details & Documents
Calibrated disposable PS inoculating loops for different applications and microbiological methods.
- Loops have an ultra-smooth surface for easy streaking
- Eliminates the need for flaming
- Aseptically manufactured in cleanroom giving a sterility assurance level 10⁻³
Specifications
- Packed:20/microseal pouch
- Capacity:5 µl
- Description:Aseptic
- Length:200 mm
- Seq:07