Inoculating loops, PS
Gamma irradiated, 10 µl, 20/microseal pouch, 200 mm
Catalog # LOCAA4G
Supplier: Local
Jump to:
Product Details & Documents
Calibrated disposable PS inoculating loops for different applications and microbiological methods.
- Loops have an ultra-smooth surface for easy streaking
- Eliminates the need for flaming
- Aseptically manufactured in cleanroom giving a sterility assurance level 10⁻³
Specifications
- Packed:20/microseal pouch
- Capacity:10 µl
- Description:Gamma irradiated
- Length:200 mm
- Seq:13