Order Entry
United Kingdom
ContactUsLinkComponent
SZ61 Trinocular with LED BASE for Transmitted (Fixed Angle) and Reflected (Variable Angle) Light - UK Power Cord Consists of: N1197700,,,,SZ61-TR,,,,Trinocular Stereo-Microscope zoom body with ESD capability, magnification range 0.67x - 4.5x, zoom ra 1 * 1 items
Catalog # OLYMK23008484
Supplier:  OLYMPUS OPTICAL
SZ61 Trinocular with LED BASE for Transmitted (Fixed Angle) and Reflected (Variable Angle) Light - UK Power Cord Consists of: N1197700,,,,SZ61-TR,,,,Trinocular Stereo-Microscope zoom body with ESD capability, magnification range 0.67x - 4.5x, zoom ra 1 * 1 items
Catalog # OLYMK23008484
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together