Order Entry
United Kingdom
ContactUsLinkComponent
SZ51 Binocular with LED BASE for Transmitted (Fixed Angle) and Reflected (Variable Angle) Light - UK Power Cord Consists of: N1197800,,,,SZ51,,,,Stereo-Microscope zoom body with ESD capability, magnification range 0.8x - 4x, zoom ratio 5:1, FN 22, in 1 * 1 items
Catalog # OLYMK23008952
Supplier:  OLYMPUS OPTICAL
SZ51 Binocular with LED BASE for Transmitted (Fixed Angle) and Reflected (Variable Angle) Light - UK Power Cord Consists of: N1197800,,,,SZ51,,,,Stereo-Microscope zoom body with ESD capability, magnification range 0.8x - 4x, zoom ratio 5:1, FN 22, in 1 * 1 items
Catalog # OLYMK23008952
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together