Order Entry
United Kingdom
ContactUsLinkComponent
Wireless (433.92MHz) 1 minute transmissions (can be customised to client needs) Range (Line-of-Sight) 100m IP68 Rated Core application Room Temperature / Ambient Standard product battery life 5 Years 1 Year Warranty 1 * 1 items
Catalog # TSCA314-001
Supplier:  T SCAN
Wireless (433.92MHz) 1 minute transmissions (can be customised to client needs) Range (Line-of-Sight) 100m IP68 Rated Core application Room Temperature / Ambient Standard product battery life 5 Years 1 Year Warranty 1 * 1 items
Catalog # TSCA314-001
Supplier:  T SCAN
Frequently Bought Together