ALEXA FLUOR® 488 AFFINIPURE FAB FRAGMENT GOAT ANTI-HUMAN IGG, FC? FRAGMENT SPECIFIC 1 * 0,5 mg
Catalog # JACK109-547-008
Supplier: Jackson Immuno Research
ALEXA FLUOR® 488 AFFINIPURE FAB FRAGMENT GOAT ANTI-HUMAN IGG, FC? FRAGMENT SPECIFIC 1 * 0,5 mg
Catalog # JACK109-547-008
Supplier: Jackson Immuno Research
Frequently Bought Together
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
GOAT FAB ANTI-MOUSE IGG (H + L) -ALEXA FLUOR 488, MINX NONE 1 * 750 µG
Supplier: Jackson Immuno Research
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Jackson Immuno Research
BIOSENSOR / ANTI-HUMAN FC CAPTURE (AHC) - Pack of 5 trays of 96 biosensors 1 * 5 items
Supplier: PALL FORTEBIO
PROCHO5 W/PLURONIC W/O L-GLN OR HT 1 * 1 L
Supplier: LONZA