KARL FISCHER REAGENT AQUAMICRON AX 1 * 500 mL
ENVACAR12
Frequently Bought Together
VWR®, Water Sampling Bottles, Sterile
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Klercide Non-Sterile Disp 100% Polyester Flat Mophead 1 * 20 items
: ECOLAB CONTAMINATION CONTROL
Karl-Fischer-Reagenz Aquamicron CXU, 10 Ampullen je 5 m
: A1 ENVIROTECH
Clinical waste container, WIVA™ Infinity
: DANIELS HEALTHCARE LIMITED