Order Entry
United Kingdom
ContactUsLinkComponent
White Crystalline Powder.Assay: 98.5-101.5% Specific rotation: +6.3 to + 7.3. Loss on drying: <0.3%.Residue on ignition: <0.3%,Chloride: <0.5% ,Sulfate: <0.03%,Iron: <0.003%,Heavy metals: <0.0015%Organic volatile impurities: passes 1 * 100 g
Catalog # ICNA0219960880
Supplier:  MP Biomedicals
White Crystalline Powder.Assay: 98.5-101.5% Specific rotation: +6.3 to + 7.3. Loss on drying: <0.3%.Residue on ignition: <0.3%,Chloride: <0.5% ,Sulfate: <0.03%,Iron: <0.003%,Heavy metals: <0.0015%Organic volatile impurities: passes 1 * 100 g
Catalog # ICNA0219960880
Supplier:  MP Biomedicals
Frequently Bought Together