Protective ointment, Travabon® classic
Protective ointment, Travabon® Classic, 1000 ml bottle
Catalog # 115-2089
Supplier: Deb-STOKO
Jump to:
Product Details & Documents
Specialist pre-work skin protection cream against stubborn, non water-soluble dirt such as oils, paints, graphite, metallic dust, adhesives and water-insoluble workplace substances. Suitable for use on hands and body. For areas where gloves are not worn.
- Facilitates cleansing of the skin thanks to soil suspending emulsifiers
- Enhanced protective action through skin protection pigments
- With glycerine and emollients
- Water-soluble, oil-free, colourant-free, silicone-free, perfumed
Ordering information: Soft bottle may only be used with Stoko® Vario® dispensers.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Specifications
- Description:Protective ointment, Travabon® Classic, 1000 ml bottle
- Seq:04