Order Entry
United Kingdom
ContactUsLinkComponent
AS950 Refrigerated Sampler, 24x1L PE, 230 VAC, AC PWR BackUp, contact LIQ DECT, STRAIN with Vinyl Tube, RS485 COM 1 * 1 items
Catalog # HACHASACXRX4141XX
Supplier:  Hach
AS950 Refrigerated Sampler, 24x1L PE, 230 VAC, AC PWR BackUp, contact LIQ DECT, STRAIN with Vinyl Tube, RS485 COM 1 * 1 items
Catalog # HACHASACXRX4141XX
Supplier:  Hach
Frequently Bought Together