Order Entry
United Kingdom
ContactUsLinkComponent
An agonist of both nAChR and mAChR receptors, with reported Ki values ranging from 10 to 10,000 nM for different receptors and diffe rent preparations 1 * 5 g
Catalog # CAYM14486-5
Supplier:  Cayman Chemical
An agonist of both nAChR and mAChR receptors, with reported Ki values ranging from 10 to 10,000 nM for different receptors and diffe rent preparations 1 * 5 g
Catalog # CAYM14486-5
Supplier:  Cayman Chemical
Frequently Bought Together