Order Entry
United Kingdom
ContactUsLinkComponent
A potent antagonist of the PAF receptor, inhibiting aggregation of rabbit and human platelets induced by PAF with IC50 values of 75 and 170 nM, respectively 1 * 1 mg
Catalog # CAYM15955-1
Supplier:  Cayman Chemical
A potent antagonist of the PAF receptor, inhibiting aggregation of rabbit and human platelets induced by PAF with IC50 values of 75 and 170 nM, respectively 1 * 1 mg
Catalog # CAYM15955-1
Supplier:  Cayman Chemical
Frequently Bought Together