Order Entry
United Kingdom
ContactUsLinkComponent
A benzylisoquinoline alkaloid that exhibits antimicrobial, anti-inflammatory, and anti-oxidant properties and demonstrates antiproli ferative and pro-apoptotic effects in some cancer cell lines 1 * 25 mg
Catalog # CAYM16951-25
Supplier:  Cayman Chemical
A benzylisoquinoline alkaloid that exhibits antimicrobial, anti-inflammatory, and anti-oxidant properties and demonstrates antiproli ferative and pro-apoptotic effects in some cancer cell lines 1 * 25 mg
Catalog # CAYM16951-25
Supplier:  Cayman Chemical
Frequently Bought Together