An acetylated form of L-carnitine that facilitates the uptake of acetyl CoA into mitochondria during fatty acid oxidation, enhances acetylcholine production, and stimulates protein and membrane phospholipid synthesis 1 * 25 g
Catalog # CAYM16948-25
Supplier: Cayman Chemical
Frequently Bought Together