Order Entry
United Kingdom
ContactUsLinkComponent
Quantitative Filter Paper, N°131, 150 mm, folded 1 * 100 items
Catalog # BINZ146007
Supplier:  Ahlstrom-Munksjö
undefined
Quantitative Filter Paper, N°131, 150 mm, folded 1 * 100 items
Catalog # BINZ146007
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together