Cleaning cloths, Unitex®
Turquoise, 380×300 mm, 200 (4×50)/pack; 156/carton
Catalog # ZVGA10148-01
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
These Unitex® cleaning cloths are excellent for cleaning and degreasing surfaces, in conjunction with cleaning agents like silicone removers, water based cleaners or water. Features special manufacturing process in which polyester and pulp fibres are woven together by hydro power.
- Tear resistant
- High strength
Specifications
- Packed:200 (4×50)/pack; 156/carton
- Colour:Turquoise
- L×W:380×300 mm
- Seq:01