: YMC-Triart C18 VS (Set of 3) 12nm, S-3um, 5 x 0.5 mm ID, 1/16 Art-Nr. TA12S03-E5J0AU 1 * 3 items
Catalog # YMCATA12S03-E5J0AU
Supplier: YMC
: YMC-Triart C18 VS (Set of 3) 12nm, S-3um, 5 x 0.5 mm ID, 1/16 Art-Nr. TA12S03-E5J0AU 1 * 3 items
Catalog # YMCATA12S03-E5J0AU
Supplier: YMC
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK