Order Entry
United Kingdom
ContactUsLinkComponent
SDF-1 α and SDF-1 β, members of the chemokine α subfamily that lack the ELR domain, were initially identified using the signa l sequence trap cloning strategy from a mouse bone-marrow stromal cell line. SDF-1 α and SDF-1 β cDNAs encode precurs 1 * 25 µG
Catalog # GENSZ03306-25
Supplier:  Genscript
SDF-1 α and SDF-1 β, members of the chemokine α subfamily that lack the ELR domain, were initially identified using the signa l sequence trap cloning strategy from a mouse bone-marrow stromal cell line. SDF-1 α and SDF-1 β cDNAs encode precurs 1 * 25 µG
Catalog # GENSZ03306-25
Supplier:  Genscript
Frequently Bought Together