Order Entry
United Kingdom
ContactUsLinkComponent
Chemokine (C-X-C motif) ligand 1 (CXCL1) is a small cytokine belonging to the CXC chemokine family that was previously called GRO1 oncogene, GRO-α, KC, neutrophil-activating protein 3 (NAP-3) and melanoma growth stimulating activity, alpha (M 1 * 25 µG
Catalog # GENSZ03303-25
Supplier:  Genscript
Chemokine (C-X-C motif) ligand 1 (CXCL1) is a small cytokine belonging to the CXC chemokine family that was previously called GRO1 oncogene, GRO-α, KC, neutrophil-activating protein 3 (NAP-3) and melanoma growth stimulating activity, alpha (M 1 * 25 µG
Catalog # GENSZ03303-25
Supplier:  Genscript
Frequently Bought Together