Interchangeable blocks for TS-DW Thermoshakers
Block for TS-DW, B-2S, One Sarstedt® 96/2200 µl deep well plate
Catalog # 460-0327
Supplier: GRANT INSTRUMENTS
Jump to:
Product Details & Documents
Specifications
- Description:Block for TS-DW, B-2S
- Seq:03
- For:One Sarstedt® 96/2200 µl deep well plate