Ringer's solution (quarter strength) standard, unbuffered
500 mL
Supplier: Thermo Fisher Scientific
Ringer's solution (quarter strength) standard, unbuffered
Catalog # J67524.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Contains 147 mM sodium chloride, 4 mM potassium chloride, and 3 mM calcium chloride in endotoxin free water and sterile filtered through 0.22 micron filter.
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pk:500 mL



