Fluorescent cell counters, Cellometer® X
Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green) and PC laptop, X2
NEXCCMX2-SK150
The Cellometer X are PC-based cell imagers for fluorescent image capture and analysis.
- Cell count and viability in less than 30 seconds
- Ability to perform cell-based assays
- Cell size range 0,5 to 15 µm and sample volume 20 µl
Both can perform cell counting, viability, intensity and population analysis, for yeast concentration in brewing fermentations.
Delivery information: Supplied with the Cellometer® software.
Documents
- Model:X2
- Description:Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green) and PC laptop
- Seq:003
- Cat. No.:NEXCCMX2-SK150
- Sample volume:20 µl
- Cell size:0,5 − 15 µm
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- NEXCCCBM-011-2MLValidation solution for Cellometer® fluorescent instruments (100 tests) in a vial1 * 2 mL - £619.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



