Order Entry
United Kingdom
ContactUsLinkComponent
10mm Quartz semi micro cell, stopper (800µl working volume) 1 * 1 items
Catalog # WAPR80-2002-81
Supplier:  Biochrom
10mm Quartz semi micro cell, stopper (800µl working volume) 1 * 1 items
Catalog # WAPR80-2002-81
Supplier:  Biochrom
Frequently Bought Together