N'-Hydroxy-2,2-dimethylpropanimidamide 95%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
N'-Hydroxy-2,2-dimethylpropanimidamide 95%
Catalog # ACRO439660010
Supplier: Thermo Fisher Scientific
CAS Number: 42956-75-2
Jump to:
Product Details & Documents
Synonyms: N-Hydroxypivalimidamide, (1Z)-N'-hydroxy-2,2-dimethylpropanimidamide, Propanimidamide, N'-hydroxy-2,2-dimethyl-
- CAS Number:42956-75-2
- Formula:C₅H₁₂N₂O
- MDL Number:MFCD03426250
- Molecular Weight:116.16 g/mol
- Melting Pt:108.6…113.7 °C
- Storage Temperature:Ambient
Documents
ACRO43966
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



