2-Iodophenol 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:533-58-4
- Formula:C₆H₅IO
- MDL Number:MFCD00013963
- Molecular Weight:220.01 g/mol
- Merck Index:13,05057
- Boiling Pt:186…187 °C (160 mmHg)
- Melting Pt:37…43 °C
- Density:1,8757 g/cm³ (80 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:208-569-9
- ADR:6.1,III
Documents
ACRO41248
SDS
Specifications
Specification Test Results
- Appearance:White to brown-red to brown Crystalline powder or low melting mass
- Infrared spectrum:Conforms
- Melting point:37 to 43 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



