Sodium methanesulphinate 85%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
Sodium methanesulphinate 85%
Catalog # ACRO392960500
Supplier: Thermo Fisher Scientific
CAS Number: 20277-69-4
Jump to:
Product Details & Documents
Synonyms: Methanesulphinic acid, sodium salt
- CAS Number:20277-69-4
- Formula:CH₃NaO₂S
- MDL Number:MFCD00040392
- Molecular Weight:102.09 g/mol
- Melting Pt:222…226 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥80 %
Specifications
- Pack type:Glass bottle
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



