Ammonium iodide ACS
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
Ammonium iodide ACS
Catalog # ACRO193212500
Supplier: Thermo Fisher Scientific
CAS Number: 12027-06-4
Jump to:
Product Details & Documents
- CAS Number:12027-06-4
- Formula:NH₄I
- MDL Number:MFCD00011424
- Molecular Weight:144.94 g/mol
- Melting Pt:551 °C (subl.)
- Density:2.51 g/cm³
- Storage Temperature:Ambient
Documents
ACRO19321
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals (May turn yellow on storage)
- Residue after ignition:≤0.05 %
- Titration:≥99.0 %
- Chloride and Bromide:≤0.005 % (as Cl)
- Phosphate (PO4):≤0.001 %
- Sulfate (SO4):≤0.05 %
- Barium (Ba):≤0.002 %
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤0.001 %
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



