Benzil 99+%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,2-Diphenyl-1,2-ethanedione
- CAS Number:134-81-6
- Formula:(C₆H₅)C(O)C(O)(C₆H₅)
- MDL Number:MFCD00003080
- Molecular Weight:210.23 g/mol
- Merck Index:13,01078
- Boiling Pt:346…348 °C (760 mmHg)
- Melting Pt:94…97 °C
- Density:1,23 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-157-0
Specifications
Specification Test Results
- Appearance:Yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:93 to 97 °C
- GC:≥99.0 %
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤500 ppm
- Potassium (K):≤200 ppm
- Sodium (Na):≤500 ppm
- Carbon compounds (as C):≤0.01 %
- Insoluble matter:≤0.02 % (in dilute HCl)
- Nitrogen compounds (as N):≤0.002 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



