9-Fluorenone 99+%
Plastic bottle, 2,5 kg
: 9H-Fluoren-9-one
- :486-25-9
- :C₁₃H₈O
- :MFCD00001141
- :180.21 g/mol
- :342 °C (760 mmHg)
- :81…85 °C
- :0,9 g/cm³ (20 °C)
- :163 °C
- :Ambient
- :207-630-7
- :3077
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:81 to 85 °C
- GC:≥99.0 %
- NMR (1H-NMR):Consistent with structure
- Pack type:Plastic bottle
- Pk:2,5 kg



