2,6-Xylidine 99%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline
- CAS Number:87-62-7
- Formula:C₈H₁₁N
- MDL Number:MFCD00007747
- Molecular Weight:121.18 g/mol
- Merck Index:12,10217
- Boiling Pt:216 °C (760 mmHg)
- Melting Pt:10…12 °C
- Density:0.98 g/cm³
- Flash Pt:91 °C
- Storage Temperature:Ambient
- EINECS:201-758-7
- UN:1711
- ADR:6.1,II
Specifications
Specification Test Results
- Appearance:Clear yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5591 to 1.5611 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with HClO4:98.5 to 101.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



