Hypoxanthine 99.5%
Glass bottle, 100 g
: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- :68-94-0
- :C₅H₄N₄O
- :MFCD00005725
- :136.11 g/mol
- :13,04895
- :>250 °C (dec.)
- :Ambient
- :200-697-3
Documents
ACRO12201
SDS
- Appearance:White to light yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.2 %
- HPLC:≥99.4 %
- Pack type:Glass bottle
- Pk:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



