Cyclohexanone oxime 97%
Plastic bottle, 2,5 kg
- :100-64-1
- :C₆H₁₁NO
- :MFCD00001660
- :113.16 g/mol
- :206 °C (1013 hPa)
- :88…92 °C
- :1,13 g/cm³ (20 °C)
- :90 °C
- :Ambient
- :202-874-0
- :1325
- Appearance:Off-white to beige to light brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- Titration with HClO4:≥96.0 %
- Pack type:Plastic bottle
- Pk:2,5 kg



