ω-Bromo-3-nitroacetophenone 97%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
ω-Bromo-3-nitroacetophenone 97%
Catalog # ACRO137470050
Supplier: Thermo Fisher Scientific
CAS Number: 2227-64-7
Jump to:
Product Details & Documents
Synonyms: 3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone
- CAS Number:2227-64-7
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00024512
- Molecular Weight:244.04 g/mol
- Melting Pt:90…94 °C
- Storage Temperature:Ambient
- EINECS:218-764-0
- UN:3261
- ADR:8,II
Documents
ACRO13747
SDS
Specifications
Specification Test Results
- Appearance:Yellow-green to beige to light orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:90 to 94 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



