Automated cell counters, Cellometer®
Cell counter with embedded computer and 10" touch screen, Auto 1000
734-2774
The Cellometer® Auto 1000 is designed for mammalian cells to perform density analysis, size measurement, Trypan Blue assays, analysis of clusters and irregular cells. A large touch screen enables the visualisation of cell morphology and selects saved assays with optimised parameters.
- Compact Cellometer® Mini with a small footprint
- Powerful software linked to the equipment via USB
- Brightfield imaging and pattern-recognition software
- Individual live cells and dead cells stained with Trypan Blue
- Cell counting, concentration, diameter and viability
- Calculation and report in less than ten seconds
The Cellometer® Auto 2000 is a tactile instrument with two fluorescence channels suitable for counting labelled cells.
Fluorescence assays can be performed in less than one minute.
Cell size range from 5 to 300 microns (CHT4-PD300-002 slides are required for cells greater than 80 microns Ø).
- Model:Auto 1000
- Description:Cell counter with embedded computer and 10" touch screen
- Seq:02
- Cat. No.:734-2774
- Cell size:5 − 60 μm
Accessories
- Description
- Description
- NEXCCHT4-PD300-003Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts1 * 500 items
- Description
- Description
- Description
- Description
- Description
- Description
- NEXCCMAUTO1K-IQOQCounting chambers reference bead solution and validation protocol compatible with Auto 10001 * 1 items
- Description
- Description
- Description
- Description
- NEXCCS2-0108-5MLViaStain™ propidium iodide (PI) staining solution for dead nucleated cells1 * 5 mL
- Description
- NEXCCS2-0106-5MLViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)1 * 1 items - £227.00
- Description
- NEXCCS2-0107-5MLViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml)1 * 1 items
- Description



