Fluorescent cell counters, Cellometer® X
Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green), X2
Catalog # NEXCMX2
Supplier: NEXCELOM BIOSCIENCE
Jump to:
Product Details & Documents
The Cellometer X are PC-based cell imagers for fluorescent image capture and analysis.
- Cell count and viability in less than 30 seconds
- Ability to perform cell-based assays
- Cell size range 0,5 to 15 µm and sample volume 20 µl
Both can perform cell counting, viability, intensity and population analysis, for yeast concentration in brewing fermentations.
Delivery information: Supplied with the Cellometer® software.
Documents
Specifications
- Model:X2
- Description:Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green)
- Seq:002
- Cat. No.:NEXCMX2
- Sample volume:20 µl
- Cell size:0,5 − 15 µm
- Measurement time:30 s
Accessories
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambersDisposable plastic counting chambersNEXCCHT4-PD100-002Log in for availability1 * 100 items - £523.00
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Laptop computer for Cellometer®
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Validation solution for Cellometer® fluorescent instruments (100 tests) in a vialValidation solution for Cellometer® fluorescent instruments (100 tests) in a vialNEXCCCBM-011-2MLLog in for availability1 * 2 mL - £619.00



