Software for Cellometer®
Software license for GMP/GLP support (single user copy)
NEXCVSL-01
The FCS Express flow cytometry analysis software enables the upgrade of a Cellometer automated cell counter to an image cytometer.
- Colourized scatter plots and histograms
- Accurate gating for enhanced sensitivity
- Import and creation of data layouts for new assays
- Convenient reports to Excel format
- Ability to perform cell based assays
- Description:Software license for GMP/GLP support (single user copy)