Counting chambers, Cellometer®
Counting chambers with protective film
Catalog # 734-2887
Supplier: NEXCELOM BIOSCIENCE
Jump to:
Product Details & Documents
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
- Two independent enclosed chambers
- Compatible with all Cellometer® cell counters
- Cell suspension (20 μl) loaded via a a single channel pipette
Specifications
- Description:Counting chambers with protective film
- Seq:02