Software for Cellometer®
Instrument upgrade for Cellometer® Vision 300
Catalog # NEXCMSYSUG110
Supplier: NEXCELOM BIOSCIENCE
Jump to:
Product Details & Documents
The FCS Express flow cytometry analysis software enables the upgrade of a Cellometer automated cell counter to an image cytometer.
- Colourized scatter plots and histograms
- Accurate gating for enhanced sensitivity
- Import and creation of data layouts for new assays
- Convenient reports to Excel format
- Ability to perform cell based assays
Documents
Specifications
- Description:Instrument upgrade for Cellometer® Vision 300