Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Microscope stand, precision, Dino-Lite, RK-10A
Microscope stand, precision, Dino-Lite, RK-10A
Vertical arm extension, RK-10-VX, length: 15 cm
Catalog # IDCPRK-10-VX
Microscope stand, precision, Dino-Lite, RK-10A
Catalog # IDCPRK-10-VX
Microscope stand, precision, Dino-Lite, RK-10A
Microscope stand, precision, Dino-Lite, RK-10A
Vertical arm extension, RK-10-VX, length: 15 cm
Price
£25.80
Log in for availability
1 * 1 items
Product Details & Documents
The Dino-Lite RK-10A is a sturdy and stable high-end stand that can be used with all professional Dino-Lite models. It is constructed of resilient stainless steel and lightweight aluminium and offers a very precise fine-focus adjustment as well as a quick release function.
  • Quick release button allows for fast and convenient vertical movement
  • Fine-focus adjustment knob is especially useful at high magnification
  • Adjustable safety stop to prevent the microscope from touching the specimen
  • Adjustable horizontal arm (RK-10-EX, included) has a 15 cm range with 360-degree rotation
  • Additional arm increases the effective range to observe larger objects
Extensions:
RK-10-EX: Additional horizontal arm (included with stand)
RK-10-VX: Vertical arm extension
RK-10-PX: XY positioning arm with accurate, smooth adjustment options, designed for all sorts of applications that require fine examination with a X- and Y-axis movement.
Delivery information: Supplied with HD-P1 holder and RK-10-EX horizontal arm.
Specifications
  • Description:
    Vertical arm extension, RK-10-VX, length: 15 cm
  • Seq:
    01