QuEChERS dSPE tubes, Q-sep™
Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black and 50 mg C18, 2 ml
Catalog # REST26219
Supplier: Restek
Jump to:
Product Details & Documents
QuEChERS dSPE tubes are used for extract cleanup. It is fast, simple sample Prep for multiresidue pesticide analysis. It has a advantage that no glassware required and it is ready to use. It can be individually labelled for easy sorbent identification and the tubes are preweighed with ultra pure sorbents.
Support original unbuffered, AOAC (2007.01), european (EN 15662) and mini-multiresidue QuEChERS methods.
Packaging: Packaged in foil subpacks of 10 for enhanced protection and storage stability.
Specifications
- Capacity:2 ml
- Description:Tubes with 150 mg magnesium sulphate, 50 mg PSA, 50 mg graphitised carbon black and 50 mg C18
- Seq:08