Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Isocyanates singles
Isocyanates singles
1,6-HDIP, 1000 µg/ml in dimethyl sulfoxide, 1 ml/ampoule, 1 Ampoul
Catalog # REST33002
Supplier:   Restek
Isocyanates singles
Catalog # REST33002
Supplier:  Restek
CAS Number:  
Isocyanates singles
Isocyanates singles
1,6-HDIP, 1000 µg/ml in dimethyl sulfoxide, 1 ml/ampoule, 1 Ampoul
CAS Number:  
Price
£84.40
Log in for availability
1 * 1 Ampoul
Product Details & Documents
OSHA 47 is for 4,4'-methylenediphenyl isocyanate (4,4'-MDI) in indoor air, and NIOSH Method 5522 is for 2,4-TDI; 2,6-TDI; 4,4'-MDI; and 1,6-hexamethylene diisocyanate (1,6-HDI) in air. Restek offers the 1,-(2-pyridyl)piperzine (1-2pp) derivative.
ASTM D5836 and OSHA 42 are test methods for determining 2,4-toluene diisocyanate (2,4-TDI) and 2,6-TDI in the workplace atmosphere.
Specifications
Specifications
  • Pk:
    1 Ampoul
  • Description:
    1,6-HDIP, 1000 µg/ml in dimethyl sulfoxide
  • Pk Info:
    1 ml/ampoule