Isocyanates singles
2,6-TDIP, 1000 µg/ml in dimethyl sulfoxide, 1 ml/ampoule, 1 Ampoul
Catalog # REST33000
Supplier: Restek
Jump to:
Product Details & Documents
OSHA 47 is for 4,4'-methylenediphenyl isocyanate (4,4'-MDI) in indoor air, and NIOSH Method 5522 is for 2,4-TDI; 2,6-TDI; 4,4'-MDI; and 1,6-hexamethylene diisocyanate (1,6-HDI) in air. Restek offers the 1,-(2-pyridyl)piperzine (1-2pp) derivative.
ASTM D5836 and OSHA 42 are test methods for determining 2,4-toluene diisocyanate (2,4-TDI) and 2,6-TDI in the workplace atmosphere.
Specifications
Specifications
- Pk:1 Ampoul
- Description:2,6-TDIP, 1000 µg/ml in dimethyl sulfoxide
- Pk Info:1 ml/ampoule