Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Volumetric flasks, class A

Volumetric flasks, class A

1000 ml, 24/20, 0,40 ± ml
Catalog # POULRB258/WAC/K/2
Volumetric flasks, class A
Catalog # POULRB258/WAC/K/2
Supplier:  
Volumetric flasks, class A
Volumetric flasks, class A

1000 ml, 24/20, 0,40 ± ml

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Volumetric flask, BLAUBRAND®, A, DE-M, 1000 ml, Boro 3.3, NS 24/29, PP stopper, ISO individual certificate 1 * 1 items Catalog #612-3626

Product 612-3626 is similar to POULRB258/WAC/K/2 but manufacturer and specifications may vary.

Volumetric flask class a 1000ml ns 24/29 with pp stopper, with serial number and individual work certificate 1 * 1 items Catalog #612-6710

Product 612-6710 is similar to POULRB258/WAC/K/2 but manufacturer and specifications may vary.

Volumetric flask BLAUBRAND® class A, DE-M marking, 1000 ml, Boro 3.3, NS 24/29, PP-stopper 1 * 1 items Catalog #612-5082

Product 612-5082 is similar to POULRB258/WAC/K/2 but manufacturer and specifications may vary.

Product Details & Documents

Borosilicate glass, with PP NS-stopper.

Individually numbered.

Certifications: DIN/BS/ISO 1042

Documents

Specifications
  • NS:
    24/20
  • Capacity:
    1000 ml
  • Seq:
    16
  • Tolerance:
    0,40 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.