IN SITU BLOCK FOR PEQSTAR X THERMOCYCLER Flat block including buffer reservoir for up to 4 glass slides. Max. heating and cooling rate: 4 C/s Block uniformity (72 C): 0.20 C Temperature range block: 4 C - 105 C 1 * 1 items
Catalog # PEQL95-051FLAT
Supplier: VWR Peqlab
Related Information
Brochure_peqSTAR_EN Broschüre_peqSTAR_DE peqSTAR Thermocycler
Product Literature
peqSTAR Thermocycler
Videos
Frequently Bought Together