Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Brightfield cell counters, Cellometer®
Brightfield cell counters, Cellometer®
Brightfield cell counter, Auto T4 Plus
Catalog # NEXCCMAUTOT4PLUS
Supplier:   NEXCELOM BIOSCIENCE
Brightfield cell counters, Cellometer®
Catalog # NEXCCMAUTOT4PLUS
Brightfield cell counters, Cellometer®
Brightfield cell counters, Cellometer®
Brightfield cell counter, Auto T4 Plus
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These automated cell counters are designed for counting small cells.
  • Highly reproducible results can be saved and exported in Excel
  • Improved data quality, versatility and flexibility
  • Templates for hundreds of cell types and easy definition of types of cells
  • Brightfield technique, powerful optics, pointer and analysis software
Applications usually include insect, algae, yeast, cancer, blood cells and spores.
Delivery information: Supplied with a box of counting chambers.
Specifications
  • Model:
    Auto T4 Plus
  • Description:
    Brightfield cell counter
  • Cat. No.:
    NEXCCMAUTOT4PLUS
  • Cell size:
    > 5 µm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2886
    Log in for availability
    1 * 75 items - £516.00
Product Information
  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    NEXCCHT4-PD100-002
    Log in for availability
    1 * 100 items - £523.00
Product Information
  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2884
    Log in for availability
    1 * 1.500 items
Product Information
  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2885
    Log in for availability
    1 * 2.500 items - £10,400.00
Product Information
  • Catalog #
    Availability
    Price
  • Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    NEXCB05-02-050
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    Availability
    Price
  • Counting chambers reference bead solution and validation protocol compatible with Auto T4
    Counting chambers reference bead solution and validation protocol compatible with Auto T4
    NEXCCMAUTOT4-IQOQ
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    Availability
    Price
  • Laptop computer for Cellometer®
    Laptop computer for Cellometer®
    NEXCCMCOMPSK150
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2887
    Log in for availability
    1 * 900 items - £2,930.00
Product Information
  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2902
    Log in for availability
    1 * 4.500 items
Product Information
  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2883
    Log in for availability
    1 * 500 items - £2,310.00
Product Information
  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    NEXCCHT4-SD100-314
    Log in for availability
    1 * 2.700 items - £7,220.00