Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Automated cell counters, Cellometer®

Automated cell counters, Cellometer®

Cell counter with small footprint controlled by external PC, Mini
Catalog # 734-2776
Supplier:   NEXCELOM BIOSCIENCE
Automated cell counters, Cellometer®
Catalog # 734-2776
Automated cell counters, Cellometer®
Automated cell counters, Cellometer®

Cell counter with small footprint controlled by external PC, Mini

Price
£3,630.00
Log in for availability
1 * 1 items
Product Details & Documents

The Cellometer® Auto 1000 is designed for mammalian cells to perform density analysis, size measurement, Trypan Blue assays, analysis of clusters and irregular cells. A large touch screen enables the visualisation of cell morphology and selects saved assays with optimised parameters.

  • Compact Cellometer® Mini with a small footprint
  • Powerful software linked to the equipment via USB
  • Brightfield imaging and pattern-recognition software
  • Individual live cells and dead cells stained with Trypan Blue
  • Cell counting, concentration, diameter and viability
  • Calculation and report in less than ten seconds

The Cellometer® Auto 2000 is a tactile instrument with two fluorescence channels suitable for counting labelled cells.

Fluorescence assays can be performed in less than one minute.

Cell size range from 5 to 300 microns (CHT4-PD300-002 slides are required for cells greater than 80 microns Ø).

Specifications
  • Model:
    Mini
  • Description:
    Cell counter with small footprint controlled by external PC
  • Seq:
    03
  • Cat. No.:
    734-2776
  • Cell size:
    5 − 60 μm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2883
    Log in for availability
    1 * 500 items - £2,310.00

Product Information

  • Catalog #
    Availability
    Price
  • Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
    Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
    NEXCCHT4-PD300-003
    Log in for availability
    1 * 500 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2902
    Log in for availability
    1 * 4.500 items

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    NEXCCHT4-PD100-002
    Log in for availability
    1 * 100 items - £523.00

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2884
    Log in for availability
    1 * 1.500 items

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2885
    Log in for availability
    1 * 2.500 items - £10,400.00

Product Information

  • Catalog #
    Availability
    Price
  • Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    NEXCB05-02-050
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers reference bead solution and validation protocol compatible with Auto 1000
    Counting chambers reference bead solution and validation protocol compatible with Auto 1000
    NEXCCMAUTO1K-IQOQ
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Laptop computer for Cellometer®
    Laptop computer for Cellometer®
    NEXCCMCOMPSK150
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2886
    Log in for availability
    1 * 75 items - £516.00

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2887
    Log in for availability
    1 * 900 items - £2,930.00

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ propidium iodide (PI) staining solution for dead nucleated cells
    ViaStain™ propidium iodide (PI) staining solution for dead nucleated cells
    NEXCCS2-0108-5ML
    Log in for availability
    1 * 5 mL

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
    ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
    NEXCCS2-0106-5ML
    Log in for availability
    1 * 1 items - £227.00

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml)
    ViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml)
    NEXCCS2-0107-5ML
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    NEXCCHT4-SD100-314
    Log in for availability
    1 * 2.700 items - £7,220.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.