Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Accessories for Cellometer®
Accessories for Cellometer®
ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
Catalog # NEXCCS2-0106-5ML
Supplier:   NEXCELOM BIOSCIENCE
Accessories for Cellometer®
Catalog # NEXCCS2-0106-5ML
Accessories for Cellometer®
Accessories for Cellometer®
ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
Price
£227.00
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
  • Seq:
    913