Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Automated cell counters, Cellometer®

Automated cell counters, Cellometer®

Cell counter with embedded computer and 10" touch screen, Auto 2000
Catalog # 734-2775
Supplier:   NEXCELOM BIOSCIENCE
Automated cell counters, Cellometer®
Catalog # 734-2775
Automated cell counters, Cellometer®
Automated cell counters, Cellometer®

Cell counter with embedded computer and 10" touch screen, Auto 2000

Price
£24,890.00
Log in for availability
1 * 1 items
Product Details & Documents

The Cellometer® Auto 1000 is designed for mammalian cells to perform density analysis, size measurement, Trypan Blue assays, analysis of clusters and irregular cells. A large touch screen enables the visualisation of cell morphology and selects saved assays with optimised parameters.

  • Compact Cellometer® Mini with a small footprint
  • Powerful software linked to the equipment via USB
  • Brightfield imaging and pattern-recognition software
  • Individual live cells and dead cells stained with Trypan Blue
  • Cell counting, concentration, diameter and viability
  • Calculation and report in less than ten seconds

The Cellometer® Auto 2000 is a tactile instrument with two fluorescence channels suitable for counting labelled cells.

Fluorescence assays can be performed in less than one minute.

Cell size range from 5 to 300 microns (CHT4-PD300-002 slides are required for cells greater than 80 microns Ø).

Specifications
  • Model:
    Auto 2000
  • Description:
    Cell counter with embedded computer and 10" touch screen
  • Seq:
    05
  • Cat. No.:
    734-2775
  • Cell size:
    5 − 60 μm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2883
    Log in for availability
    1 * 500 items - £2,310.00

Product Information

  • Catalog #
    Availability
    Price
  • Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
    Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
    NEXCCHT4-PD300-003
    Log in for availability
    1 * 500 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2902
    Log in for availability
    1 * 4.500 items

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    NEXCCHT4-PD100-002
    Log in for availability
    1 * 100 items - £523.00

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2884
    Log in for availability
    1 * 1.500 items

Product Information

  • Catalog #
    Availability
    Price
  • Disposable plastic counting chambers
    Disposable plastic counting chambers
    734-2885
    Log in for availability
    1 * 2.500 items - £10,400.00

Product Information

  • Catalog #
    Availability
    Price
  • Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    Polystyrene beads (Ø = 5 µm) in Trypan blue solution (5×106 beads/ml)
    NEXCB05-02-050
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers reference bead solution and validation protocol compatible with Auto 1000
    Counting chambers reference bead solution and validation protocol compatible with Auto 1000
    NEXCCMAUTO1K-IQOQ
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Laptop computer for Cellometer®
    Laptop computer for Cellometer®
    NEXCCMCOMPSK150
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2886
    Log in for availability
    1 * 75 items - £516.00

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    734-2887
    Log in for availability
    1 * 900 items - £2,930.00

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ propidium iodide (PI) staining solution for dead nucleated cells
    ViaStain™ propidium iodide (PI) staining solution for dead nucleated cells
    NEXCCS2-0108-5ML
    Log in for availability
    1 * 5 mL

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
    ViaStain™ acridine orange (AO) and propidium iodide (PI) staining solution for live and dead mammalian nucleated cells (5 ml)
    NEXCCS2-0106-5ML
    Log in for availability
    1 * 1 items - £227.00

Product Information

  • Catalog #
    Availability
    Price
  • ViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml)
    ViaStain™ acridine orange (AO) staining solution for nucleated cells (5 ml)
    NEXCCS2-0107-5ML
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Counting chambers with protective film
    Counting chambers with protective film
    NEXCCHT4-SD100-314
    Log in for availability
    1 * 2.700 items - £7,220.00