Order Entry
United Kingdom
ContactUsLinkComponent
Stainless Steel Rack with 7 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high with high fronts and lip pulls Each tray has a card holder on the front Each tray holds 4 boxes Each rack holds 28 boxes NB U360 holds 3 racks per shelf 1 * 1 items
Catalog # AVIAUN194107HLF/LH
Supplier:  AVIAMAX
Stainless Steel Rack with 7 trays Rack hold boxes up to 134 x 134 x 55mm The trays are 10mm high with high fronts and lip pulls Each tray has a card holder on the front Each tray holds 4 boxes Each rack holds 28 boxes NB U360 holds 3 racks per shelf 1 * 1 items
Catalog # AVIAUN194107HLF/LH
Supplier:  AVIAMAX
Frequently Bought Together